siRNA Details
| Gene Symbol: Oosp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037634.3 | NP_001032723.1 | 4 | 463 | ||
| GenBank mRNA | AK143411.1 | - | 4 | 464 | ||
| BC099498.1 | - | 4 | 423 | 30931566 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oosp2| Gene Symbol: Oosp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037634.3 | NP_001032723.1 | 4 | 463 | ||
| GenBank mRNA | AK143411.1 | - | 4 | 464 | ||
| BC099498.1 | - | 4 | 423 | 30931566 | ||