siRNA Details
| Gene Symbol: Ercc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011729.2 | NP_035859.2 | 3 | 664 | ||
| GenBank mRNA | AK038822.1 | - | 3 | 493 | ||
| AK137563.1 | - | 3 | 496 | |||
| AK140825.1 | - | 3 | 493 | |||
| AK160645.1 | - | 1 | 36 | |||
| AK164205.1 | - | 3 | 482 | |||
| BC033413.1 | - | 3 | 384 | |||
| BC053034.1 | - | 3 | 473 | |||
| BC060659.1 | - | 3 | 463 | |||
| BC092235.1 | - | 3 | 455 | |||
| BC141070.1 | - | 3 | 494 | 9055682 | ||
| D16306.1 | - | 3 | 359 | |||
| U40796.1 | - | 3 | 560 | |||

Ercc5
Genomic Map