siRNA Details
| Gene Symbol: Trpm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153417.1 | NP_700466.1 | 26 | 3719 | ||
| GenBank mRNA | AY135644.1 | - | 26 | 3719 | ||
| BC022929.1 | - | 6 | 900 | |||
| BC145223.1 | - | 26 | 3671 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trpm6| Gene Symbol: Trpm6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153417.1 | NP_700466.1 | 26 | 3719 | ||
| GenBank mRNA | AY135644.1 | - | 26 | 3719 | ||
| BC022929.1 | - | 6 | 900 | |||
| BC145223.1 | - | 26 | 3671 | |||