siRNA Details
| Gene Symbol: Rorb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001043354.2 | NP_001036819.1 | 4 | 1203 | ||
| NM_001289921.1 | NP_001276850.1 | 5 | 722 | |||
| NM_146095.4 | NP_666207.3 | 4 | 669 | |||
| GenBank mRNA | AB258405.1 | - | 4 | 830 | ||
| AK044421.1 | - | 4 | 664 | |||
| AK134790.1 | - | 4 | 970 | |||
| BC024768.1 | - | 5 | 705 | |||
| BC024842.1 | - | 5 | 722 | 5358124 | ||
| BC058269.1 | - | 3 | 576 | |||
| DQ779923.1 | - | 3 | 573 | |||
| DQ779924.1 | - | 4 | 598 | |||

Rorb
Genomic Map