siRNA Details
| Gene Symbol: Rorb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001043354.2 | NP_001036819.1 | 8 | 1653 | ||
| NM_001289921.1 | NP_001276850.1 | 9 | 1172 | |||
| NM_146095.4 | NP_666207.3 | 8 | 1119 | |||
| GenBank mRNA | AB258405.1 | - | 8 | 1280 | ||
| AK134790.1 | - | 8 | 1420 | |||
| BC024842.1 | - | 9 | 1172 | 5358124 | ||
| BC058269.1 | - | 7 | 1026 | |||
| DQ779923.1 | - | 7 | 1023 | |||
| DQ779924.1 | - | 8 | 1048 | |||

Rorb
Genomic Map