siRNA Details
| Gene Symbol: Smc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252684.1 | NP_001239613.1 | 18 | 2540 | ||
| NM_001252685.1 | NP_001239614.1 | 18 | 2369 | |||
| NM_153808.2 | NP_722503.1 | 18 | 2540 | |||
| GenBank mRNA | AJ534941.1 | - | 18 | 2461 | ||
| AK122321.1 | - | 11 | 1486 | |||
| AK161254.1 | - | 18 | 2370 | |||
| BC007174.1 | - | 9 | 1096 | 3493165 | ||
| BC038345.1 | - | 18 | 2520 | 3596405 | ||

Smc5
Genomic Map