siRNA Details
| Gene Symbol: Hectd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163471.1 | NP_001156943.1 | 13 | 1512 | ||
| NM_172637.3 | NP_766225.2 | 13 | 1509 | |||
| GenBank mRNA | AK029549.1 | - | 13 | 1369 | ||
| AK142777.1 | - | 11 | 1327 | |||
| BC137671.1 | - | 13 | 1483 | 8860690 | ||
| BC144815.1 | - | 13 | 1483 | 9053363 | ||

Hectd2
Genomic Map