siRNA Details
| Gene Symbol: Yes1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205132.1 | NP_001192061.1 | 10 | 1399 | ||
| NM_001205133.1 | NP_001192062.1 | 10 | 1399 | |||
| NM_009535.3 | NP_033561.1 | 10 | 1861 | |||
| GenBank mRNA | AK052478.1 | - | 10 | 1351 | ||
| AK167420.1 | - | 10 | 1536 | |||
| BC010594.1 | - | 10 | 1235 | 3989783 | ||
| L25762.1 | - | 1 | 22 | |||
| X67677.1 | - | 10 | 1788 | |||

Yes1
Genomic Map