siRNA Details
| Gene Symbol: Erlin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164359.1 | NP_001157831.1 | 9, 10 | 853 | ||
| NM_001164360.1 | NP_001157832.1 | 9, 10 | 761 | |||
| NM_145502.3 | NP_663477.3 | 8, 9 | 957 | |||
| GenBank mRNA | AK133557.1 | - | 8, 9 | 700 | ||
| AK145990.1 | - | 9, 10 | 734 | |||
| BC011220.1 | - | 9, 10 | 725 | 4208238 | ||
| BC023849.1 | - | 8, 9 | 820 | 5345801 | ||

Erlin1
Genomic Map