siRNA Details
Gene Symbol: Peo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153796.3 | NP_722491.2 | 1 | 1304 | ||
GenBank mRNA | AK146244.1 | - | 1 | 1198 | ||
AY059385.1 | - | 1 | 766 | |||
BC071195.1 | - | 1 | 1304 | 5100486 |
