siRNA Details
| Gene Symbol: Tmem185b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146103.2 | NP_666215.1 | 1 | 1195 | ||
| GenBank mRNA | AK031130.1 | - | 1 | 1191 | ||
| BC020408.1 | - | 1 | 1183 | |||
| BC024712.1 | - | 1 | 1182 | 2646072 | ||
| BC051523.1 | - | 1 | 52 | |||

Tmem185b
Genomic Map