siRNA Details
| Gene Symbol: Ipo9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153774.1 | NP_722469.1 | 5, 6 | 661 | ||
| GenBank mRNA | AF273672.1 | - | 5, 6 | 606 | ||
| AF273673.1 | - | 5, 6 | 606 | |||
| AJ309238.1 | - | 5, 6 | 600 | |||
| AK003047.1 | - | 5, 6 | 471 | |||
| AK036687.1 | - | 5, 6 | 611 | |||
| AK173127.1 | - | 5, 6 | 601 | |||
| BC056469.1 | - | 5, 6 | 628 | |||
| BC079546.1 | - | 6, 7 | 772 | |||
| BC098508.1 | - | 5, 6 | 611 | 30015513 | ||

Ipo9
Genomic Map