siRNA Details
| Gene Symbol: Zfp101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009542.2 | NP_033568.2 | 4 | 355 | ||
| GenBank mRNA | AK076355.1 | - | 4 | 359 | ||
| AK078458.1 | - | 4 | 343 | |||
| AK169234.1 | - | 4 | 356 | |||
| U07861.1 | - | 4 | 315 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp101| Gene Symbol: Zfp101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009542.2 | NP_033568.2 | 4 | 355 | ||
| GenBank mRNA | AK076355.1 | - | 4 | 359 | ||
| AK078458.1 | - | 4 | 343 | |||
| AK169234.1 | - | 4 | 356 | |||
| U07861.1 | - | 4 | 315 | |||