siRNA Details
| Gene Symbol: Rnf103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146898.1 | - | 4 | 2498 | ||
| AK168644.1 | - | 4 | 2679 | |||
| BC137624.1 | - | 5 | 2036 | 8860643 | ||
| D76445.1 | - | 4 | 2510 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf103| Gene Symbol: Rnf103 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK146898.1 | - | 4 | 2498 | ||
| AK168644.1 | - | 4 | 2679 | |||
| BC137624.1 | - | 5 | 2036 | 8860643 | ||
| D76445.1 | - | 4 | 2510 | |||