siRNA Details
| Gene Symbol: Zfp105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009544.2 | NP_033570.2 | 4 | 816 | ||
| GenBank mRNA | AF045565.1 | - | 4 | 806 | ||
| AK147198.1 | - | 4 | 823 | |||
| BC052165.1 | - | 4 | 816 | 6515711 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp105| Gene Symbol: Zfp105 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009544.2 | NP_033570.2 | 4 | 816 | ||
| GenBank mRNA | AF045565.1 | - | 4 | 806 | ||
| AK147198.1 | - | 4 | 823 | |||
| BC052165.1 | - | 4 | 816 | 6515711 | ||