siRNA Details
| Gene Symbol: Zbtb41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172643.5 | NP_766231.2 | 2 | 1121 | ||
| GenBank mRNA | AK034756.1 | - | 1 | 220 | ||
| AK036544.1 | - | 2 | 1120 | |||
| AK163302.1 | - | 1 | 394 | |||
| BC046465.1 | - | 1 | 290 | |||
| BC150859.1 | - | 1 | 460 | 9087770 | ||
| BC172012.1 | - | 1 | 460 | |||

Zbtb41
Genomic Map