siRNA Details
| Gene Symbol: Gm4846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164306.1 | NP_001157778.1 | 1 | 154 | ||
| GenBank mRNA | BC147800.1 | - | 1 | 154 | 9087517 | |
| BC147804.1 | - | 1 | 162 | 9087521 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm4846| Gene Symbol: Gm4846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164306.1 | NP_001157778.1 | 1 | 154 | ||
| GenBank mRNA | BC147800.1 | - | 1 | 154 | 9087517 | |
| BC147804.1 | - | 1 | 162 | 9087521 | ||