siRNA Details
| Gene Symbol: Zfp148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011749.4 | NP_035879.1 | 9 | 2797 | ||
| GenBank mRNA | AF316548.1 | - | 9 | 2736 | ||
| AF316549.1 | - | 1 | 897 | |||
| AK020783.1 | - | 1 | 43 | |||
| AK036543.1 | - | 1 | 507 | |||
| AK146591.1 | - | 9 | 2659 | |||
| BC026144.1 | - | 9 | 2662 | 3991738 | ||
| U80078.1 | - | 6 | 2321 | |||
| X98096.1 | - | 9 | 2694 | |||

Zfp148
Genomic Map