siRNA Details
| Gene Symbol: Zfp148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011749.4 | NP_035879.1 | 9 | 1919 | ||
| GenBank mRNA | AF316548.1 | - | 9 | 1858 | ||
| AF316549.1 | - | 1 | 19 | |||
| AF316550.1 | - | 1 | 637 | |||
| AK036835.1 | - | 6 | 1192 | |||
| AK146591.1 | - | 9 | 1781 | |||
| BC026144.1 | - | 9 | 1784 | 3991738 | ||
| U80078.1 | - | 6 | 1443 | |||
| X98096.1 | - | 9 | 1816 | |||

Zfp148
Genomic Map