siRNA Details
| Gene Symbol: Mark1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145515.2 | NP_663490.2 | 15 | 1785 | ||
| GenBank mRNA | AB185282.1 | - | 15 | 1708 | ||
| AF453686.1 | - | 15 | 1715 | |||
| AK173181.1 | - | 14 | 1635 | |||
| BC112400.1 | - | 15 | 1714 | 40059725 | ||

Mark1
Genomic Map