siRNA Details
| Gene Symbol: Mark1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145515.2 | NP_663490.2 | 13 | 1412 | ||
| GenBank mRNA | AB185282.1 | - | 13 | 1335 | ||
| AF453686.1 | - | 13 | 1342 | |||
| AK173181.1 | - | 12 | 1262 | |||
| BC112400.1 | - | 13 | 1341 | 40059725 | ||
| CX210969.1 | - | 5 | 472 | |||

Mark1
Genomic Map