siRNA Details
| Gene Symbol: Zfp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001044697.2 | NP_001038162.1 | 6 | 618 | ||
| NM_001044698.2 | NP_001038163.1 | 6 | 614 | |||
| NM_001044700.2 | NP_001038165.1 | 6 | 570 | |||
| NM_001294323.1 | NP_001281252.1 | 6 | 577 | |||
| NM_178447.3 | NP_848542.1 | 7 | 655 | |||
| GenBank mRNA | AF483520.1 | - | 1 | 18 | ||
| AF483521.1 | - | 1 | 18 | |||
| AK036776.1 | - | 7 | 645 | |||
| AK131978.1 | - | 6 | 564 | |||
| AK141471.1 | - | 6 | 594 | |||
| BC052029.1 | - | 4 | 318 | |||
| BC057171.1 | - | 6 | 584 | |||
| BC089506.1 | - | 6 | 549 | 30247022 | ||
| CJ044064.1 | - | 6 | 563 | |||
| Y00850.1 | - | 4 | 450 | |||

Zfp2
Genomic Map