siRNA Details
| Gene Symbol: Vash2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144879.2 | NP_659128.2 | 6 | 1216 | ||
| GenBank mRNA | AK045261.1 | - | 6 | 1012 | ||
| AK078596.1 | - | 6 | 1085 | |||
| AK140989.1 | - | 6 | 1210 | |||
| AK163193.1 | - | 6 | 992 | |||
| BC024141.1 | - | 6 | 986 | 5149750 | ||

Vash2
Genomic Map