siRNA Details
Gene Symbol: Vash2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144879.2 | NP_659128.2 | 6, 7 | 1294 | ||
GenBank mRNA | AK045261.1 | - | 6, 7 | 1090 | ||
AK078596.1 | - | 6, 7 | 1163 | |||
AK140989.1 | - | 6, 7 | 1288 | |||
AK163193.1 | - | 6, 7 | 1070 | |||
BC024141.1 | - | 6, 7 | 1064 | 5149750 |
