siRNA Details
| Gene Symbol: Hhat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144881.4 | NP_659130.2 | 12 | 1765 | ||
| GenBank mRNA | AK028230.1 | - | 12 | 1568 | ||
| AK172126.1 | - | 12 | 1612 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hhat| Gene Symbol: Hhat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144881.4 | NP_659130.2 | 12 | 1765 | ||
| GenBank mRNA | AK028230.1 | - | 12 | 1568 | ||
| AK172126.1 | - | 12 | 1612 | |||