siRNA Details
| Gene Symbol: Sbspon | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033288.3 | NP_001028460.1 | 5 | 843 | ||
| GenBank mRNA | AK137144.1 | - | 5 | 844 | ||
| AK140569.1 | - | 2434 | ||||
| AK143251.1 | - | 5 | 817 | |||
| BC147194.1 | - | 5 | 830 | 9056240 | ||
| BC147195.1 | - | 5 | 830 | 9056224 | ||

Sbspon
Genomic Map