siRNA Details
| Gene Symbol: Zpr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011752.2 | NP_035882.1 | 11 | 1195 | ||
| GenBank mRNA | AB041612.1 | - | 11 | 1078 | ||
| AK016551.1 | - | 5 | 362 | |||
| AK044472.1 | - | 11 | 1084 | |||
| AK158995.1 | - | 11 | 1156 | |||
| AK167849.1 | - | 11 | 1066 | |||
| AK168061.1 | - | 11 | 1037 | |||
| AK194385.1 | - | 2 | 156 | |||
| BC021397.1 | - | 11 | 1078 | 5067274 | ||

Zpr1
Genomic Map