siRNA Details
| Gene Symbol: Zfp35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011755.2 | NP_035885.2 | 3 | 664 | ||
| GenBank mRNA | AK132703.1 | - | 3 | 667 | ||
| BC138830.1 | - | 3 | 583 | 8861852 | ||
| BC138832.1 | - | 2 | 581 | 8861854 | ||
| X17617.1 | - | 3 | 615 | |||

Zfp35
Genomic Map