siRNA Details
| Gene Symbol: Arhgef4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183019.2 | NP_898840.2 | 5, 6 | 1054 | ||
| GenBank mRNA | AK034013.1 | - | 5, 6 | 1018 | ||
| AK122448.2 | - | 1, 2 | 109 | |||
| AK134629.1 | - | 1017 | ||||
| AK138598.1 | - | 5, 6 | 1014 | |||
| AK163424.1 | - | 5, 6 | 1036 | |||
| AK169957.1 | - | 5, 6 | 1074 | |||
| BC055804.1 | - | 5, 6 | 1015 | |||
| BC094608.1 | - | 4574 | ||||
| BC139371.1 | - | 5, 6 | 1037 | 8862393 | ||
| BC139397.1 | - | 5, 6 | 1037 | 8862419 | ||

Arhgef4
Genomic Map