siRNA Details
| Gene Symbol: Zfp39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011758.2 | NP_035888.2 | 5 | 758 | ||
| GenBank mRNA | AK045944.1 | - | 5 | 759 | ||
| AK141135.1 | - | 5 | 747 | |||
| BC011182.1 | - | 1 | 21 | |||
| BC108271.1 | - | 5 | 698 | 30102085 | ||

Zfp39
Genomic Map