siRNA Details
| Gene Symbol: Slc9a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033289.2 | NP_001028461.2 | 2 | 543 | ||
| GenBank mRNA | AK077026.1 | - | 2 | 635 | ||
| AK138779.1 | - | 2 | 920 | |||
| AK166114.1 | - | 2 | 659 | |||
| BC103685.1 | - | 2 | 543 | 40048593 | ||
| BC104737.1 | - | 2 | 543 | 40048591 | ||
| BC105676.1 | - | 2 | 542 | |||

Slc9a2
Genomic Map