siRNA Details
| Gene Symbol: Hibch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146108.1 | NP_666220.1 | 7 | 511 | ||
| GenBank mRNA | AK076038.1 | - | 7 | 510 | ||
| AK082965.1 | - | 7 | 510 | |||
| AK186066.1 | - | 4 | 184 | |||
| BC026437.1 | - | 7 | 511 | 4238681 | ||

Hibch
Genomic Map