siRNA Details
Gene Symbol: Zfp57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013745.2 | NP_001013767.1 | 5 | 781 | ||
NM_001168501.1 | NP_001161973.1 | 5 | 647 | |||
NM_001168502.1 | NP_001161974.1 | 6 | 1023 | |||
GenBank mRNA | AK054083.1 | - | 5 | 764 | ||
AK135881.1 | - | 5 | 755 | |||
AK136241.1 | - | 5 | 765 | |||
AK139625.1 | - | 6 | 1023 | |||
AK143118.1 | - | 3 | 1629 | |||
AK145368.1 | - | 5 | 753 | |||
BC052028.1 | - | 3 | 948 | 5686931 | ||
D21850.1 | - | 4 | 776 |
