siRNA Details
| Gene Symbol: Abca12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175210.3 | NP_780419.2 | 37 | 5908 | ||
| GenBank mRNA | BC158053.1 | - | 37 | 5721 | 9088162 | |
| BC158063.1 | - | 37 | 5721 | 9007362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abca12| Gene Symbol: Abca12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175210.3 | NP_780419.2 | 37 | 5908 | ||
| GenBank mRNA | BC158053.1 | - | 37 | 5721 | 9088162 | |
| BC158063.1 | - | 37 | 5721 | 9007362 | ||