siRNA Details
| Gene Symbol: Fam134a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_170755.2 | NP_739561.2 | 6 | 734 | ||
| GenBank mRNA | AK034667.1 | - | 6 | 735 | ||
| AK152268.1 | - | 6 | 449 | |||
| AK158940.1 | - | 6 | 504 | |||
| AK160400.1 | - | 6 | 688 | |||
| AK165957.1 | - | 6 | 713 | |||
| AK172093.1 | - | 6 | 731 | |||
| AK185264.1 | - | 227 | ||||
| AK200447.1 | - | 2 | 59 | |||
| BC026519.1 | - | 2 | 85 | |||
| BC038286.1 | - | 2 | 120 | 4221441 | ||
| BC070406.1 | - | 6 | 726 | 5707319 | ||

Fam134a
Genomic Map