siRNA Details
| Gene Symbol: Dgkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177646.3 | NP_808314.2 | 6 | 673 | ||
| GenBank mRNA | AK172901.1 | - | 2 | 118 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dgkd| Gene Symbol: Dgkd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177646.3 | NP_808314.2 | 6 | 673 | ||
| GenBank mRNA | AK172901.1 | - | 2 | 118 | ||