siRNA Details
| Gene Symbol: Farp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145519.2 | NP_663494.2 | 3, 4 | 400 | ||
| GenBank mRNA | AK050860.1 | - | 3, 4 | 360 | ||
| AK086699.1 | - | 3, 4 | 393 | |||
| AK171713.1 | - | 3, 4 | 426 | |||
| BB663505.1 | - | 3, 4 | 393 | |||
| BC009153.1 | - | 3, 4 | 389 | 2655209 | ||
| BI739824.1 | - | 1, 2 | 25 | |||

Farp2
Genomic Map