siRNA Details
| Gene Symbol: Farp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145519.2 | NP_663494.2 | 15 | 1721 | ||
| GenBank mRNA | AK050860.1 | - | 15 | 1681 | ||
| AK171713.1 | - | 15 | 1747 | |||
| BC009153.1 | - | 15 | 1710 | 2655209 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Farp2| Gene Symbol: Farp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145519.2 | NP_663494.2 | 15 | 1721 | ||
| GenBank mRNA | AK050860.1 | - | 15 | 1681 | ||
| AK171713.1 | - | 15 | 1747 | |||
| BC009153.1 | - | 15 | 1710 | 2655209 | ||