siRNA Details
| Gene Symbol: Zfp94 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199321.1 | NP_001186250.1 | 5, 6 | 498 | ||
| NM_009568.3 | NP_033594.1 | 4, 5 | 459 | |||
| GenBank mRNA | AK138205.1 | - | 5, 6 | 439 | ||
| AK164334.1 | - | 5, 6 | 451 | |||
| BC005437.1 | - | 4, 5 | 459 | 3591835 | ||
| BC095945.1 | - | 3, 4 | 279 | 4936327 | ||
| CJ174209.1 | - | 4, 5 | 361 | |||
| U46187.1 | - | 4, 5 | 402 | |||

Zfp94
Genomic Map