siRNA Details
Gene Symbol: Zscan12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016684.2 | NP_057893.2 | 2 | 248 | ||
GenBank mRNA | AK075886.1 | - | 2 | 242 | ||
AK129138.1 | - | 2 | 248 | |||
AK139837.1 | - | 3 | 571 | |||
AK142005.1 | - | 2 | 227 | |||
BC027041.1 | - | 1 | 206 | 5364568 | ||
U62908.1 | - | 1 | 557 |
