siRNA Details
| Gene Symbol: Zscan12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016684.2 | NP_057893.2 | 2 | 218 | ||
| GenBank mRNA | AK075886.1 | - | 2 | 212 | ||
| AK129138.1 | - | 2 | 218 | |||
| AK139837.1 | - | 3 | 541 | |||
| AK142005.1 | - | 2 | 197 | |||
| BC027041.1 | - | 1 | 176 | 5364568 | ||
| U62908.1 | - | 1 | 527 | |||

Zscan12
Genomic Map