siRNA Details
| Gene Symbol: Tor4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146115.4 | NP_666227.2 | 2 | 913 | ||
| GenBank mRNA | AK043559.1 | - | 2 | 714 | ||
| AK082899.1 | - | 2 | 714 | |||
| AK153600.1 | - | 1 | 413 | |||
| AK154508.1 | - | 2 | 714 | |||
| AK155540.1 | - | 3 | 874 | |||
| AK171551.1 | - | 2 | 761 | |||
| AK172142.1 | - | 2 | 738 | |||
| BC013092.1 | - | 2 | 826 | 3584061 | ||

Tor4a
Genomic Map