siRNA Details
| Gene Symbol: Zfpm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011766.5 | NP_035896.1 | 8 | 3223 | ||
| GenBank mRNA | AF107306.1 | - | 8 | 2744 | ||
| AF118845.1 | - | 8 | 2993 | |||
| AF125166.1 | - | 8 | 3224 | |||
| BC059241.1 | - | 9 | 3069 | 6408975 | ||
| EF514219.1 | - | 8 | 4046 | |||

Zfpm2
Genomic Map