siRNA Details
| Gene Symbol: Zfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011767.2 | NP_035897.2 | 20 | 3225 | ||
| GenBank mRNA | AF071059.1 | - | 20 | 3223 | ||
| AK011329.1 | - | 5 | 545 | |||
| AK034963.1 | - | 13 | 1794 | |||
| AK041429.1 | - | 4 | 478 | |||
| AK158938.1 | - | 12 | 1533 | |||
| BC006962.1 | - | 10 | 1272 | |||
| BC094220.1 | - | 19 | 3112 | |||
| BC141139.1 | - | 20 | 3209 | 9055751 | ||

Zfr
Genomic Map