siRNA Details
| Gene Symbol: Ddx31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033294.3 | NP_001028466.2 | 9 | 1098 | ||
| GenBank mRNA | AK030888.1 | - | 9 | 1045 | ||
| AK144739.1 | - | 9 | 811 | |||
| AK166327.1 | - | 9 | 768 | |||
| BC066017.1 | - | 9 | 735 | 6401415 | ||
| BC132351.1 | - | 9 | 788 | 40130628 | ||

Ddx31
Genomic Map