siRNA Details
| Gene Symbol: Nup188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198304.2 | NP_938046.2 | 32 | 3486 | ||
| GenBank mRNA | AK129073.1 | - | 32 | 3486 | ||
| BC032090.1 | - | 32 | 3488 | |||
| BC056489.1 | - | 18 | 2065 | |||
| BC096591.1 | - | 32 | 3487 | 4916413 | ||

Nup188
Genomic Map