siRNA Details
| Gene Symbol: Nup188 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198304.2 | NP_938046.2 | 34 | 3730 | ||
| GenBank mRNA | AK082836.1 | - | 1 | 73 | ||
| AK129073.1 | - | 34 | 3730 | |||
| BC025526.1 | - | 1 | 24 | |||
| BC032090.1 | - | 34 | 3732 | |||
| BC056489.1 | - | 20 | 2309 | |||
| BC096591.1 | - | 34 | 3731 | 4916413 | ||

Nup188
Genomic Map