siRNA Details
| Gene Symbol: Pip5kl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198191.2 | NP_937834.1 | 9 | 939 | ||
| GenBank mRNA | AK133244.1 | - | 9 | 2347 | ||
| AK164998.1 | - | 9 | 1042 | |||
| AY376879.1 | - | 9 | 978 | |||
| BC028795.1 | - | 2 | 251 | |||
| BC053963.1 | - | 2 | 260 | |||
| BC117018.1 | - | 9 | 931 | 40126337 | ||
| BC119037.1 | - | 9 | 931 | 8733786 | ||

Pip5kl1
Genomic Map