siRNA Details
| Gene Symbol: Lrsam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199302.2 | NP_955006.1 | 3 | 335 | ||
| GenBank mRNA | BC049146.1 | - | 3 | 331 | 6306042 | |
| CX237785.1 | - | 4 | 370 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrsam1| Gene Symbol: Lrsam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199302.2 | NP_955006.1 | 3 | 335 | ||
| GenBank mRNA | BC049146.1 | - | 3 | 331 | 6306042 | |
| CX237785.1 | - | 4 | 370 | |||