siRNA Details
| Gene Symbol: Rabepk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145522.4 | NP_663497.2 | 3 | 312 | ||
| GenBank mRNA | AK035343.1 | - | 3 | 192 | ||
| AK153022.1 | - | 3 | 216 | |||
| BC019800.1 | - | 3 | 188 | 5150243 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rabepk| Gene Symbol: Rabepk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145522.4 | NP_663497.2 | 3 | 312 | ||
| GenBank mRNA | AK035343.1 | - | 3 | 192 | ||
| AK153022.1 | - | 3 | 216 | |||
| BC019800.1 | - | 3 | 188 | 5150243 | ||